Would you like to log in again?
Sklarcut™ Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sklarcut™
15-3569 15-3567 15-3571
76192-022EA 305.35 USD InStock
76192-022 10015-662 76192-024
Sklarcut™ Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar®
Scissors
Sklarcut™ Metzenbaum-Lahey Dissecting Scissors are used for dissecting and cutting tissue only.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
These scissors are not recommended for cutting sutures or drains. Metzenbaum-Lahey scissors tend to be shorter than standard Metzenbaum scissors. Sklarcut™ scissors feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue. These serrations assist in securely holding the tissue during cutting and minimizes tissue from slipping so the most accurate and clean cuts can be made. Sklarcut™ scissors are identified by a single gold coated finger ring on the handle of the serrated blade.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty