Would you like to log in again?
Supercut Sklarhone™ Solz 'Gold Tip' Scissors, OR Grade, Sklar
Supplier: Sklar
Supercut Sklarhone™
15-9301 15-9302 15-9303
76192-034EA 961.91 USD InStock
76192-034 76192-032 76192-030
Supercut Sklarhone™ Solz 'Gold Tip' Scissors, OR Grade, Sklar
Scissors
Sklar's® Solz gold tip scissors, also know as Kaye or Sims, are a dissecting scissor used in plastic surgery procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The design of the shank facilitates ideal atraumatic seperation of the tissue layers. Supercut Sklarhone™ scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone™ scissors minimize tearing of tissue so cuts and edges are smooth and clean.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty