Would you like to log in again?
Sklar Edge™ Tungsten Carbide Mayo Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sklar Edge™
16-1525 16-1515 16-1520 16-1625 16-1615 16-3531 16-3530 16-3533 16-1605 16-1505 16-3532 16-1620
76192-044EA 1214.94 USD InStock
76192-044 76192-066 89025-674 76192-042 76192-040 89025-678 89025-668 76192-072 76192-070 76192-038 89025-672 76192-068
Sklar Edge™ Tungsten Carbide Mayo Dissecting Scissors, OR Grade, Sklar®
Scissors
Sklar Edge™ TC Mayo Dissecting Scissors are a modification of standard Mayo scissors that provide an exceptionally hard and durable cutting edge designed to stay sharp for a significantly longer period of time than standard scissors
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar permanently bonds the tungsten carbide inserts to the blade of the scissors which maximizes control and precision as well. Mayo Scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Identified by the handle of two gold coated finger rings.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty