Would you like to log in again?
Sklar Edge™ Tungsten Carbide Supercut Sklarhone™ Iris Scissors, OR Grade, Sklar®
Supplier: Sklar
Sklar Edge™
16-3545 16-3546
76192-078EA 481.89 USD InStock
76192-078 76192-080
Sklar Edge™ Tungsten Carbide Supercut Sklarhone™ Iris Scissors, OR Grade, Sklar®
Scissors
Sklar Edge™ TC Supercut Sklarhone™ Iris scissors provide effortless, precise cutting.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
These scissors feature one knife-like razor edge and one micro-serrated edge along with the strength and durability of tungsten carbide. They can be used to cut both delicate and heavy tissue with minimal tearing or pulling. Sklar permanently bonds tungsten carbide inserts to the blades, which maximizes control and precision. These scissors are also designed to stay sharp for a significantly longer period of time than standard scissors. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty