Would you like to log in again?
Sklar Edge™ Tungsten Carbide Supercut Sklarhone™ Mayo-Stille Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
Sklar Edge™
16-3587 16-3577 16-3572 16-3582
76192-088EA 481.89 USD InStock
76192-088 76192-086 76192-084 76192-082
Sklar Edge™ Tungsten Carbide Supercut Sklarhone™ Mayo-Stille Dissecting Scissors, OR Grade, Sklar®
Scissors
Sklar Edge™ Tungsten Carbide Supercut Sklarhone™ Mayo-Stille Dissecting Scissors provide effortless, precise cutting with the benefit of the strength and durability of tungsten carbide.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
These scissors feature one knife-like razor edge and one micro-serrated scissors edge and can be used to cut both delicate and heavy tissue with minimal tearing or pulling. Sklar Edge™ Tungsten Carbide Scissors provide an exceptionally hard and durable cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than other non tungsten carbide scissors. This instrument is differentiated from other scissors by it's handle of one black finger ring and one gold finger ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty