Would you like to log in again?
Cairns Hemostatic and Scalp Forceps, OR Grade, Sklar
Supplier: Sklar
17-3041 17-3040
76192-094EA 72.68 USD InStock
76192-094 76192-092
Cairns Hemostatic and Scalp Forceps, OR Grade, Sklar
Forceps
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar's® Cairns scalp forceps are a non-ratched finger ring intracranial clamp for neurosurgical procedures. The serrated jaws ensure a firm grip, for grasping blood vessels to control bleeding. They are available curved or straight.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty