Would you like to log in again?
Sklar Edge™ Tungsten Carbide Cottle Dorsal Scissors, OR Grade, Sklar®
Supplier: Sklar
Sklar Edge™
16-3630
76192-152EA 331.97 USD InStock
76192-152
Sklar Edge™ Tungsten Carbide Cottle Dorsal Scissors, OR Grade, Sklar®
Scissors
Sklar Edge™ TC Cottle dorsal scissors are used in rhinoplasty procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The angled shanks reduce surgeon fatigue, and the curvature allows for manipulation of the soft tissue around the dorsum of the nose. Available individually, or as a component of Sklar's major rhinoplasty set. Sklar Edge™ Tungsten Carbide Scissors provide an exceptionally durable and hard cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than standard scissors. Sklar permanently bonds tungsten carbide inserts to the blades, which maximizes control and precision. Sklar Edge™ scissors can be identified by two gold coated finger rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty