Would you like to log in again?
Cushing Artery Forceps, OR Grade, Sklar®
Supplier: Sklar
17-1980
76192-208EA 67.28 USD InStock
76192-208
Cushing Artery Forceps, OR Grade, Sklar®
Forceps
Sklar's® Cushing hemostatic artery forceps are ratcheted finger ring instruments with a serrated jaw grip for control of blood flow during neurological procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Similar to the Lovelace, the forceps have straight jaws and blunt tips, but the Cushing jaw is half serrated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty