Would you like to log in again?
Duplay Dressing Forceps, OR Grade, Sklar®
Supplier: Sklar
19-1031 19-1030
76192-242EA 204.45 USD InStock
76192-242 76192-252
Duplay Dressing Forceps, OR Grade, Sklar®
Forceps
Sklar's® Duplay dressing forceps are ratcheted finger ring instruments used in nasal polyp procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
They have a serrated, fenstrated tip, and uniquely shaped long shanks. They are available with straight or curved jaw tip. The Gross-Maier forceps have a similar jaw, but straight shanks and no locking ratchet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty