Would you like to log in again?
Carroll Needle Holder, OR Grade, Sklar®
Supplier: Sklar
20-3110
76192-322EA 418.05 USD InStock
76192-322
Carroll Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® ratcheted finger ring Carroll needle holder is used for suturing.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The shorter shanks make it more suitable in plastic surgery procedures. The blunt cross-serrated jaws ensure a firm grip on the needle. While very similar to the Crile-Murray, the Carroll is distinguised by the grooved jaw.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty