Would you like to log in again?
Econo™ Multi-Cut Utility Scissors, Floor Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Econo™
21-105HG 21-100 21-105LG 21-105RB 21-105BG 21-105TB 21-105MG 21-105NB 21-105YL 21-105OR 21-105PR
76192-364BX 129.73 USD InStock
76192-364 76192-342 76192-340 76192-362 76192-338 76192-336 76192-334 76192-368 76192-332 76192-366 76192-344
Econo™ Multi-Cut Utility Scissors, Floor Grade, Sklar®
Scissors
Econo™ Utility Scissors are a multipurpose instrument designed to cut nonsterile materials such as dressings, casts, and surgical drapes.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
These scissors are straight, with one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. One plastic finger ring is larger than the other for maximized control while cutting. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty