Would you like to log in again?
TC Gillies Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-2662 21-2660 21-2664
89023-102EA 382.78 USD InStock
89023-102 76192-400 76192-346
TC Gillies Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Tungsten Carbide Gillies Needle Holder is designed for use with delicate suturing needles for closing wounds.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The device has an offset design with curved, cross-serrated tips with scissors incorporated to make it a versatile tool, including plastic surgery procedures. The Gillies needle holder has on offset larger ring. Sklar® permanently bonds tungsten carbide inserts to the instrument, ensuring they stay sharp for a longer period of time, and maximizes control and precision. TC instruments are identified by gold finger rings.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty