Would you like to log in again?
TC Thomson-Walker Bozeman Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-2175
76192-384EA 652.82 USD InStock
76192-384
TC Thomson-Walker Bozeman Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® TC Thomson-Walker Bozeman needle holder is a ratched finger ring instrument to hold curved needles while suturing.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Serrated jaws ensure a firm grip, and the curved shaft places the tip at a deeper plane. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty