Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Econo™ Vienna Nasal Speculum, Floor Grade, Sklar®
Supplier: Sklar
Econo™
21-258 21-267
76192-396BX 374.7 USD InStock
76192-396 89023-104
Econo™ Vienna Nasal Speculum, Floor Grade, Sklar®
Specula
Econo™ Vienna Nasal Speculum widens the nasal cavity for examination. It features a plier spring-handle and is available in infant, child, and adult sizes.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard, with smooth working surface
Econo™ instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. The infant size speculum features blades 25 mm long and 4 to 8 mm wide. The adult size speculum features blades 35 mm long and 5 to 10 mm wide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty