Would you like to log in again?
Mayo-Russian Tissue Forceps, OR Grade, Sklar®
Supplier: Sklar
19-2320
76192-436EA 171.26 USD InStock
76192-436
Mayo-Russian Tissue Forceps, OR Grade, Sklar®
Forceps
Sklar® Mayo-Russian Tissue Forceps are thumb forceps used to grasp dense tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar® Mayo-Russian Tissue Forceps are commonly used to hold the tissue during wound debridement and closure. Additionally, they may be used during dental procedures. The large, rounded, scoop ends have deep teeth that provide surface area and secure grip which is useful for grasping tooth fragments and placing or removing gauze.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty