Would you like to log in again?
Wright Derf Needle Holder, OR Grade, Sklar®
Supplier: Sklar
20-1549
76192-450EA 375.8 USD InStock
76192-450
Wright Derf Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Wright Derf Needle Holder features short jaws and delicate serrations that hold small needles and commonly 5-0 and 6-0 sutures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Frequently utilized in ophthalmic, dental, or plastic procedures, the shorter length allows the Derf Needle Holder to be easily controlled in smaller surgical areas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty