Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Econo™ Babcock Forceps, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-441
76192-478BX 222.4 USD InStock
76192-478
Econo™ Babcock Forceps, Floor Grade, Sklar
Forceps
Econo™ Babcock Forceps are finger ring, ratcheted, non-perforating forceps used to grasp delicate tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Econo™ Babcock Forceps are frequently used with intestinal and laparotomy procedures. Babcock Forceps are similar to Allis forceps, however they may be considered less traumatic due to the wider, rounded grasping surface. The jaws are circumferential, and the tips are triangular and fenestrated with horizontal serrations. They are particularly useful for grasping tube-shaped structures. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty