Would you like to log in again?
Econo™ Semken Dressing Forceps, Floor Grade, Sklar®
Supplier: Sklar
Econo™
21-478
76192-490BX 41.78 USD InStock
76192-490
Econo™ Semken Dressing Forceps, Floor Grade, Sklar®
Forceps
Econo™ Semken Dressing Forceps are most commonly used for application and changing of dressings.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
These forceps feature narrow tips that may be beneficial for neurosurgery or other procedures performed in constricted areas. They may also be used for dissection of fine or single-layer tissue. Serrated tips provide a solid grip on both materials and tissue. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty