Would you like to log in again?
M.G.H. Needle Holder, OR Grade, Sklar®
Supplier: Sklar
20-2730
76192-522EA 227.67 USD InStock
76192-522
M.G.H. Needle Holder, OR Grade, Sklar®
Forceps
The M.G.H. (Mass General Hospital) ratcheted finger ring needle holder is used for suturing.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Sklar® Mass General Hospital ratcheted finger ring needle holder features cross-serrated, fenstrated jaws to ensure a firm grip on the needle. It has short, straight jaws that are blunt, distinguishing them from the sharp Crile-Wood, Saro, Halsey, and Webster.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty