Would you like to log in again?
Adson Needle Holder, OR Grade, Sklar®
Supplier: Sklar
20-2740
76192-526EA 190.57 USD InStock
76192-526
Adson Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Adson Needle Holder is a ratcheted finger ring instrument used for holding needles during suturing.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
This product is straight with serrated jaws. One jaw is completely fenestrated and one jaw has a groove.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty