Would you like to log in again?
Wangensteen Needle Holder, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
20-2781 20-2780
76192-528EA 417.49 USD InStock
76192-528 76192-530
Wangensteen Needle Holder, OR Grade, Sklar®
Forceps
The Wangensteen needle nolder has pyramid-shaped teeth to reduce grip slippage when using fine needles.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The straight jaws are to be used with 1-0, 2-0, and 3-0 sutures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty