Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Econo™ Pederson Vaginal Speculum, Floor Grade, Sklar®
Supplier: Sklar
21-368 21-334
76192-538BX 387.88 USD InStock
76192-538 76192-458
Econo™ Pederson Vaginal Speculum, Floor Grade, Sklar®
Specula
The Econo™ Pederson Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, pap smears, and examining the cervix.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Econo™ Pederson Vaginal Speculum is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The blades of the Pederson Speculum are more narrow than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient. Econo™ instruments are ready-to-use, disposable, floor grade instruments sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty