Would you like to log in again?
Econo™ Ballenger Sponge Forceps, Floor Grade, Sklar®
Supplier: Sklar
21-571
76192-602BX 184.16 USD InStock
76192-602
Econo™ Ballenger Sponge Forceps, Floor Grade, Sklar®
Forceps
Econo® Ballenger Sponge Forceps are a ratcheted, finger ring instrument used to grasp tissue or hold sponges.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Econo® Ballenger Sponge Forceps may also be known as Foerster sponge forceps, however the shorter length instrument is often referred to as Ballenger. Sponge forceps may also be known as sponge sticks. The instrument is used to hold gauze or cotton that is precisely wrapped around the tips and the ratcheted is locked to secure a firm grip. This creates a sponge stick that can be used in any type of procedure to absorb blood and excess fluids from the surgical site and surrounding areas. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty