Would you like to log in again?
Econo™ Jansen Nasal Bayonet Forceps, Floor Grade, Sklar®
Supplier: Sklar
21-605
76192-604BX 159.85 USD InStock
76192-604
Econo™ Jansen Nasal Bayonet Forceps, Floor Grade, Sklar®
Forceps
Sklar® Jansen Bayonet thumb forceps are commonly used in nasal procedures for placing packing.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The bayonet style handle and long jaws allow an unobstructed view when working in the nasal cavity. Serrated tips securely hold surgical materials. These forceps may also be used in intracranial procedures. Econo™ instruments are floor grade, disposable instruments, which eliminate the need for cleaning and reprocessing. Because they are disposable, Econo™ instruments also reduce the likelihood of cross-contamination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty