Would you like to log in again?
Econo™ Gerald Tissue Forceps, Floor Grade, Sklar®
Supplier: Sklar
21-612
76192-610BX 154.19 USD InStock
76192-610
Econo™ Gerald Tissue Forceps, Floor Grade, Sklar®
Forceps
Econo™ Gerald Tissue Forceps are a light to intermediate weight instrument with very narrow tips specifically used to handle delicate tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Econo™ Gerald Tissue Forceps are often used in cardiothoracic procedures. They have a stop peg to prevent an overly harsh grasp that may crush the tissue. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty