Would you like to log in again?
Econo™ Hoff Towel Clamps, Floor Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Econo™
21-611 21-655
89023-260PK 194.76 USD InStock
89023-260 76192-618
Econo™ Hoff Towel Clamps, Floor Grade, Sklar
Forceps
Econo™ Hoff Towel Clamps are a non-perforating clamp used to secure instruments and towels to the drape surrounding a surgical site.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The jaws and tips are curved, while the shanks and handle are flat so they can rest on the patient. The tip of this towel clamp are round with a cross-serrated surface, so they grip tightly but do not penetrate the surgical drape. They have ratcheted finger ring handles, so a firm hold can be secured. This clamp should not be used on skin.
Econo™ instruments are disposable.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty