Would you like to log in again?
Econo™ Frazier Tissue Hook, OR Grade, Sklar®
Supplier: Sklar
Econo™
21-801
76192-650BX 145.3 USD InStock
76192-650
Econo™ Frazier Tissue Hook, OR Grade, Sklar®
Retractors
The Econo™ Frazier tissue hook is a single-prong tissue hook used for gentle handling and retracting of tissue, often used in dermatologic procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Econo™ Frazier tissue hook is straight with an extra delicate, sharp hook tip. Econo™ instruments are floor-grade, disposable instruments that eliminate the need for cleaning and reprocessing. Because they are disposable, Econo™ instruments also reduce the likelihood of cross-contamination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty