Would you like to log in again?
Langenbeck Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8067 21-8085 21-8083 21-8068 21-8069 21-8081
76192-680EA 682.83 USD InStock
76192-680 76192-734 76192-732 76192-678 76192-730 76192-676
Langenbeck Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Langenbeck needle holder is used for delicate and refined suturing.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Langenbeck needle holder features plier handles, but has no spring action and no locking mechanism. This instrument is often chosen by users who prefer a quick release of the needle when suturing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty