Would you like to log in again?
TC Halsey Needle Holder, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
21-8099 21-8000 21-8098 21-8001
89023-306EA 270.69 USD InStock
89023-306 89023-308 76192-692 76192-690
TC Halsey Needle Holder, OR Grade, Sklar®
Forceps
Sklar® TC Halsey Needle Holders are ratcheted, used for holding small needles and fine suture material which is commonly used in plastic surgery or subcutaneous procedures around the face, hands, and delicate tissue, such as dermatology.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
TC Halsey Needle Holders are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle, however serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty