Would you like to log in again?
TC Metzenbaum Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8101 21-8100
76192-694EA 790.65 USD InStock
76192-694 76192-696
TC Metzenbaum Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Tungsten Carbide Metzenbaum needle holder is of the spring handle plier variety.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The TC Metzenbaum needle holder is most commonly used for grasping needles for suturing in plastic surgery procedures. The curved serrated jaws ensure a strong grip. This product is similar to the Mathieu-Ryder but with a wider jaw tip. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty