Would you like to log in again?
Mathieu Delicate Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8109 21-8106
76192-704EA 297.33 USD InStock
76192-704 76192-762
Mathieu Delicate Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Mathieu Needle Holder is a single spring instrument with a plier handle grip that curves inward at the bottom and features a ratchet locking mechanism at the base of the handles that can be easily squeezed to be locked and unlocked.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
This needle holder is used when more refined suturing is required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty