Would you like to log in again?
TC Toennis Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8066
76192-728EA 352.27 USD InStock
76192-728
TC Toennis Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Tungsten Carbide Toennis Needle Holder is the plier handle variety.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Toennis needle holder is most commonly used for grasping needles for suturing in plastic surgery procedures. It is longer than the very similar Crile. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. These instruments are identified by gold handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty