Would you like to log in again?
Baby Doyen Intestinal Forceps, OR Grade, Sklar®
Supplier: Sklar
22-2154 22-2155
76192-738EA 352.53 USD InStock
76192-738 76192-740
Baby Doyen Intestinal Forceps, OR Grade, Sklar®
Forceps
Sklar® Doyen-Baby Intestinal Forceps are used to grasp and enclose delicate structures such as the intestines.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar® Doyen-Baby Intestinal Forceps have a delicate pattern with diagonal serrations. The forceps are available with curved or straight jaws and finger ring handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty