Would you like to log in again?
Goldman-Fox Scissors, OR Grade, Sklar
Supplier: Sklar
22-2519 22-2520
76192-742EA 209.76 USD InStock
76192-742 89025-286
Goldman-Fox Scissors, OR Grade, Sklar
Scissors
The Sklar® Goldman-Fox Scissors are finger ring scissors primarily used in dental procedures to trim gingival tissue, but may also be used in plastic surgery or hemorrhoidectomy and other rectal procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Goldman-Fox Scissors have beveled blades, one of which is serrated, that taper into sharp, fine tips. The serrations provide a firm grip on tissue and the tapered blades make small, precise cuts in small surgical areas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty