Would you like to log in again?
Reynolds Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
22-2770 22-2768 22-2722 22-2759 22-2758 22-2762 22-2772 22-2760
75817-768EA 221.86 USD InStock
75817-768 76192-758 76192-756 89025-308 76192-746 76192-754 76192-752 76192-750
Reynolds Dissecting Scissors, OR Grade, Sklar®
Scissors
Sklar® Reynolds Dissecting Scissors are designed to grip tissue and make a clean cut. The upper blade is sharp, which cuts the tissue, while the lower blade is serrated to grip and hold the tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty