Would you like to log in again?
Mixter Right Angle Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
22-3270 22-3257 17-1762 22-3255 18-1250 17-1772 55-2892 55-2894
76192-760EA 196.15 USD InStock
76192-760 10193-878 10197-586 10192-260 10197-584 10194-750 89025-350 89025-352
Mixter Right Angle Forceps, OR Grade, Sklar
Forceps
Sklar® Mixter Right Angle Forceps are ratcheted, finger ring forceps commonly used for working in obscured surgical sites
Mixter Right Angle Forceps are available in multiple lengths and have serrations the entire length of the jaw.
They are most frequently used for clamping, dissection, or grasping tissue.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty