Would you like to log in again?
Semken Dressing Forceps, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
21-813 22-4250 21-872 22-4154 22-4050 21-812 22-4152 22-4150 22-4156
89025-368EA 73.29 USD InStock
89025-368 76192-846 10847-254 76192-778 89025-370 76192-878 76192-776 76192-876 76192-874
Semken Dressing Forceps, OR Grade, Sklar®
Forceps
Sklar® Semken Dressing Forceps are most commonly used for application and changing of dressings.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
These forceps feature narrow tips that may be beneficial for neurosurgery or other procedures performed in constricted areas. They may also be used for dissection of fine or single-layer tissue. Serrated tips provide a solid grip on both materials and tissue.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty