Would you like to log in again?
TC Castroviejo Needle Holder with Diamond Coated Jaw, OR Grade, Sklar®
Supplier: Sklar
21-8130 21-8131
76192-782EA 1443.45 USD InStock
76192-782 76192-780
TC Castroviejo Needle Holder with Diamond Coated Jaw, OR Grade, Sklar®
Forceps
The Sklar® Castroviejo needle holders are used for micro surgery, for suturing and stapling.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Castroviejo needle holders hold very fine suture needles in opthalmic procedures. Castroviejo's can be opened from any rotational position, and are available straight or curved, with or without a lock. The diamond jaw serration prevents rotation of the needle. Sklar® permanently bonds tungsten carbide inserts to the instrument, ensuring shaprness for a longer period of time, and maximizes control and precision. The tungsten carbine line of instruments is identified by gold handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty