Would you like to log in again?
TC Jacobson Needle Holder with Diamond Coated Jaw, OR Grade, Sklar®
Supplier: Sklar
21-8156 21-8151 21-8253 21-8158 21-8160 21-8257 21-8157 21-8255 21-8159 21-8254 21-8152 21-8134 21-8150 21-8135 21-8139 21-8259 21-8138 21-8140 21-8251 21-8141 21-8133 21-8258 21-8250 21-8132
76192-808EA 1151.08 USD InStock
76192-808 76192-806 76192-828 76192-818 76192-804 76192-838 76192-816 76192-802 76192-814 76192-836 76192-800 76192-812 76192-834 76192-798 76192-842 76192-810 76192-832 76192-788 76192-796 76192-840 76192-830 76192-786 76192-784 76192-790
TC Jacobson Needle Holder with Diamond Coated Jaw, OR Grade, Sklar®
Forceps
The Sklar® Jacobson Tungsten Carbide Microvascular Needle Holder is primarily used for suturing in microvascular and cardiovascular procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Jacobson Tungsten Carbide Microvascular Needle Holder is also known as a Jacobson-Castroviejo Needle Holder. The finely cross-serrated, tapered jaws are able to securely hold very fine sutures. Serrations on TC instruments are designed to stay sharp for a significantly longer period of time than standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty