Would you like to log in again?
Wire Eye Speculum, OR Grade, Sklar®
Supplier: Sklar
22-8824 22-8828 22-8826
76192-916EA 101.53 USD InStock
76192-916 76192-914 76192-912
Wire Eye Speculum, OR Grade, Sklar®
Specula
The Sklar® wire eye speculum is an opthalmic instrument used to separate and hold open the eyelids.
  • With industry standard
  • Reusable and non-Sterile
  • Premium OR-grade
The tension in this spring wire device allows the lightweight, open wire to gently retract both the top and bottom eyelids without excessive pressure on the globe. The choice of eye speculum is typically dependent upon the anatomical nature of the patient's eyes as well as the specific procedure being performed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty