Would you like to log in again?
Parsonnet Epicardial Retractors, OR Grade, Sklar®
Supplier: Sklar
22-8847 22-8845 22-8846
76192-928EA 348.74 USD InStock
76192-928 76192-926 76192-930
Parsonnet Epicardial Retractors, OR Grade, Sklar®
Retractors
The Sklar® Parsonnet Epicardial Retractor is a tiny, self-retaining retractor commonly used in coronary bypass surgery.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Parsonnet Epicardial Retractor is available with 3x3 or 4x3 sharp prongs, and is used to retract epicardial fat and expose arteries in the cardiac muscle for anastomosis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty