Would you like to log in again?
Gregory Stay Suture Clamp Forceps, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
24-3011 24-3010 24-3013 24-3012
76192-988EA 575.19 USD InStock
76192-988 76192-986 76192-984 76192-982
Gregory Stay Suture Clamp Forceps, OR Grade, Sklar®
Forceps
Sklar® Gregory Stay Suture Clamp Forceps are small, lightweight forceps with smooth jaws that are used during microsurgical procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The small clamps allow sutures to be temporarily placed to hold the area that is being operated on. The instrument comes in a variety of lengths and curvatures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty