Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Deschamps Ligature Carrier, OR Grade, Sklar®
Supplier: Sklar
26-1685 26-1780 26-1680 26-1785
76193-010EA 257.43 USD InStock
76193-010 76193-008 76193-012 76193-014
Deschamps Ligature Carrier, OR Grade, Sklar®
Sutures
The Sklar® Dechamps Ligature Carriers are used for guiding suture material into difficult to reach areas or deep muscle and tissue.
  • Latex-free and reusable
  • Industry standard
  • Non-sterile
The Dechamps Ligature Carrier is frequently used in vaginal surgeries where the unique angle and curvature of the tip allows the user to place lateral ligatures. The tip of the instrument may be sharp or blunt, and curved for either right or left hand. Both styles are sharp enough to penetrate tough tissue, but not cause damage to surrounding vessels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty