Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Fiber-optic Macintosh Laryngoscope Blades, Sklar®
Supplier: Sklar
07-1274 07-1268 07-1273 07-1269 07-1272
76193-064EA 293.12 USD InStock
76193-064 76193-082 76193-062 76193-080 76191-592
Fiber-optic Macintosh Laryngoscope Blades, Sklar®
Scalpels and Scalpel Assemblies Scalpel Blades
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Macintosh Laryngoscope Blades are used for direct, rigid laryngoscopy procedures to view vocal cords, remove foreign objects from the throat, biopsy, or tracheal intubation. These fiber-optic blades follow the Green System of fiber-optic compatibility, which is indicated by a green bar where the blade attaches to a fiber-optic handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty