Would you like to log in again?
Sklarhone™ Metzenbaum-Nelson Bariatric Dissecting Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-3412 15-3409
76191-742EA 384.4 USD InStock
76191-742 76193-066
Sklarhone™ Metzenbaum-Nelson Bariatric Dissecting Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Supercut Sklarhone™ Metzenbaum-Nelson Bariatric Scissors are long dissecting scissors most commonly used in Bariatric Surgeries such as gastric bypass, gastric banding, and sleeve gastrectomy. They are also used in Nephrectomy, Cystectomy, and Prostatectomy procedures to dissect and cut tissue. These scissors are not recommended for cutting sutures or drains. Supercut Sklarhone™ scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone™ scissors minimize tearing of tissue so cuts and edges are smooth and clean. Sklarhone™ instruments are identified by dual black finger rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty