Would you like to log in again?
CleanMax® Clean and Sterile Disposable Coveralls with Elastic Wrist/Ankle, Lakeland Industries
Supplier: Lakeland Industries
CTL417CS-5X CTL417CS-XL CTL417CS-3X CTL417CS-MD CTL417CS-2X CTL417CS-LG CTL417CS-4X
76193-340CS 990.24 USD InStock
76193-340 76193-330 76193-328 76193-336 76193-338 76193-332 76193-334
CleanMax® Clean and Sterile Disposable Coveralls with Elastic Wrist/Ankle, Lakeland Industries
Disposable Garments
Clean and sterile disposable coveralls are individually double-bagged and folded to promote easier gowning with less excursions.
  • Clean and sterile
  • Manufactured in silicone-free environment
  • Latex-free
  • Tunneled elastic on wrists (with thumb loops), ankles, and back half of waist
  • Individually double-bagged
  • Certificate of radiation included
  • Gamma radiation indicator dots on each package
CleanMax® cleanroom garments provide comfort, quality, and protection. CleanMax® sterilized garments are both clean manufactured and sterile (not all sterile garments are clean manufactured).
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunneled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certifications: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty