Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TARBP2 Rabbit Polyclonal Antibody

Supplier: ProSci

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

33-041
76194-040EA 632.13 USD InStock
76194-040
Anti-TARBP2 Rabbit Polyclonal Antibody
Antibodies
TRBP2 (TAR (HIV-1) RNA binding protein 2) , along with Dicer and Argonaute-2, is part of the RISC (RNA induced silencing complex). As part of the RISC, TRBP2 is important in viral defense. Dicer cleaves dsRNA into siRNA. Through TRBP2-Dicer interaction, the siRNA template is loaded onto Argonaute-2, which cleaves encountered viral mRNA strands complementary to the loaded siRNA base-pair sequence.

Western blot: 5-10 ug/ml Provided assay concentrations are suggestions only, antibody titration may be required for optimal results.

Type: Primary
Antigen: TARBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Primate

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty