Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IGHM Mouse Monoclonal Antibody [clone: IM260]

Supplier: ProSci
33-147
76195-516EA 723.64 USD InStock
76195-516
Anti-IGHM Mouse Monoclonal Antibody [clone: IM260]
Antibodies
IgM is the first antibody generated in an immune response to an antigen. It is generally a pentamer with each of the five immunoglobulins linked together with disulfide bonds. In its pentamer form, it has a molecular mass of 970 kDa and 10 antigen binding sites (due to the large size of most antigens, not all binding sites can be filled simultaneously.  IgM antibodies account for approximately 5%-10% of all the antibody in the body.<p>This mAb is specific for the human IgM heavy chain. It does not cross-react with other immunoglobulin heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. IgM antibody is useful in the identification of certain cancer types. Some tumors express a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Flow Cytometry: 0.5-1ug/10e6 cells IF: 0.5-1 ug/ml WB: 0.5-1 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) Due to differences in protocols and secondaries, the anti-IgM antibody may need to be titered for optimal performance.<p>1. FFPE staining requires boiling sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.<br>2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: IGHM
Clonality: Monoclonal
Clone: IM260
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty