Would you like to log in again?
ActivArmr® 97-631 High Dexterity Cold Protection Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ActivArmr®
97631110 97631080 97631100 97631070 97631090
76196-268CS 408.19 USD InStock
76196-268 76196-286 76196-262 76196-284 76196-260
ActivArmr® 97-631 High Dexterity Cold Protection Gloves, Ansell
Gloves
Feel everything but the cold. With an industry leading combination of comfort, warmth, and dexterity, this glove is certifiably warm and helps to fight frostbite.
  • PVC coating stays more flexible than competitive set in cold temperatures
The PVC coating provides excellent grip, even in the coldest temperatures. The interior is made of a unique two layer design; flexible nylon to protect against mechanical risks, and soft, insulative acrylic to provide comfort and warmth.
Certifications: Highest possible dexterity score on EN 420 test. ANSI level 3 abrasion resistance. ANSI level A2 cut protection. EN 388:2016 - 2231B mechanical hazards. EN 511 - 12X cold protection. REACH Compliant. CE marked. Food safe. Compliant with FDA food handling requirements.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty