Would you like to log in again?
Edge® 48-703 Industrial Work Gloves with Leather Palm, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Edge®
48703070 48703110 48703090 48703080 48703100
76196-278CS 1719.36 USD InStock
76196-278 76196-294 76196-282 76196-292 76196-280
Edge® 48-703 Industrial Work Gloves with Leather Palm, Ansell
Gloves
Advanced cut and abrasion protection combined with premium leather for a superior grip.
  • Good dexterity and flexibility
  • Medium cut protection and high levels of abrasion protection
  • Good grip in dry and slightly oily environments
  • Good sensitivity, fit, and cushioning
  • Polyethylene/glass fiber liner
Made with a seamless liner as well as a knitted wrist to offer close fit in cut resistant applications. Combines a good level of cut, abrasion, and thermal resistance in one glove. The leather palm re-enforcement provides extra protection for the worker in cut and hot parts handling.
Certifications: CE 0493 marked. EN 388:2003 - 4543 mechanical hazards. EN 388:2016 - 4X43D mechanical hazards. EN 407 - X1XXXX heat protection. REACH Complaint.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty